Koolit® Sweat-Proof Gel Packs, Moisture-Resistant Gel Pack Refrigerants, Cold Chain Technologies
Fournisseur: Cold Chain Technologies
Certificats
A propos de cet article
The 800 series Koolit® ice gel packs are sweat-proof and designed to absorb condensation
The outer material is spun-woven and backed to a polyethylene film for outstanding moisture absorbance. The 0°C (32°F) CMC-based ice gel is non-toxic and ideal for use in the shipment of refrigerated products.
Customers Who Bought This Also Bought