Dipper, High Density Polyethylene, Dynalon
Fournisseur: Dynalon
Certificats
A propos de cet article
Manufactured from chemically resistant HDPE
Dipper for removing samples from large tanks. Features a PVC hand grip. Dippers feature two pour spouts and are graduated. Product is lightweight but extremely durable.
Ordering information: Additional sizes are available upon request.
Customers Who Bought This Also Bought