VWR® Balance Curved Forceps
Catalog # 89259-950
Supplier: VWR International
Specifications
- Length:102 mm (4")
- Material:Stainless steel
- Cat. no.:89259-950
Specifications
About this item
VWR® Balance Forceps are curved with plastic tips.
- Stainless steel construction
Curved forceps are designed for precise handling of fine objects, built from non-corrosive stainless steel and featuring inert plastic tips.
Customers Who Bought This Also Bought