VWR® Lister Bandage Scissors
# de catalogue 89259-978
Fournisseur: VWR International
Spécifications
- Material:Stainless steel
- Cat. no.:89259-978
- Length:140 mm (51/2")
Spécifications
A propos de cet article
VWR® Lister Bandage Scissors are stainless steel with one blade with a rounded tip and one blade with a blunt, flattened, probe tip.
- Scissors are angular
Lister bandage scissors are available in a range of lengths and order sizes, from individual to bulk.
Customers Who Bought This Also Bought