VWR® Student Forceps
Catalog # 89259-940
Supplier: VWR International
Specifications
- Length:127 mm (5")
- Material:Stainless steel
- Cat. no.:89259-940
Specifications
About this item
VWR® Student Forceps are stainless steel forceps with blunt serrated tips.
- Blunt serrated tips
- Stainless steel construction
- Durable for years of student use
Serrated tips help to ensure firm grasp of objects without slippage.
Frequently Bought Together