VWR® Silicone Smart Cap Gaskets
Fournisseur: VWR International
Certificats
A propos de cet article
VWR® USP Class VI Platinum-Cured Silicone gaskets come in a replacement 5-pack, and are designed to fit perfectly with Versatile Caps.
- Fully autoclavable
- Designed to fit Versatile Caps
Customers Who Bought This Also Bought