HyperSep Dispersive SPE Mylar™ Pouches, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Convenient and effective approach for determining pesticide residues in fruit, vegetables and other foods.
QuEChERS reagents are provided in individual pouches
Ordering information: Each pack contains 50 pouches (no centrifuge tubes are supplied).
Customers Who Bought This Also Bought