HyperSep Dispersive SPE Multipacks, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Convenient and effective approach for determining pesticide residues in fruit, vegetables and other foods.
QuEChERS reagents are provided in individual metalized pouches
Ordering information: Each pack of 50 pouches is supplied with 50 empty centrifuge tubes with plug seal caps.
Customers Who Bought This Also Bought