Nitro™ and Tattoo™ Earplugs, 3M™
# de catalogue CA12000-181
Fournisseur: 3M
Spécifications
- NRR:32 dB
- Color:Assorted
- Description:Uncorded earplugs with Tattoo™ design
- Cat. no.:CA12000-181
- Packaging:Poly bag
- Supplier No.:P1000
Spécifications
A propos de cet article
Nitro™ and Tattoo™ Earplugs feature bright colors and cutting edge graphics with a comfortable, tapered design for all-day wear with great protection.
Customers Who Bought This Also Bought