EVAHL PAPRs (Powered Air Purifying Respirators), Blower Assemblies, Bullard®
Fournisseur: Bullard
Certificats
A propos de cet article
The EVAHL is simple, safe, and dependable. The system features one-button operation, a choice of two air speeds, and a single filter design.
- Certified for hazardous locations (Class I Division 2, Class II Division 2, Class III)
- Versatile respirator for particulate or chemical environments
- Lightweight, compact, well-balanced design
Additionally, the EVAHL offers Active Flow Technology™, which maintains an airflow of 8.5 CFM (high speed), 1000 Assigned Protection Factor (depending upon head top), and dual alarms for low flow and low battery. Finally, the EVAHL has a visual battery fuel gauge to check remaining charge at the push of a button, a brushless motor with ten times the life of a brush motor PAPR, and optional genius charger for battery trending analysis.
Certifications: NIOSH Approvals TC-21C-0922 and TC-23C-2765. ANSI/ISA-12.12.01-2007 (CSA Certificate 2510928) Class I and II, Division and Class III, Division 1 and 2 Hazardous (Classifed) Location.
Ordering information: Filters/cartridges must be ordered separately (except for EVAHL4, where a single HE filter is included).
Spécifications
- Catalog No:
- 89402-900
- 89402-898
- 89402-896
- 89402-894
- Airflow:
- 8.5/7.0 cfm (Hi/Lo Speed)
- 8.5/7.0 cfm (Hi/Lo Speed)
- 8.5/7.0 cfm (Hi/Lo Speed)
- 8.5/7.0 cfm (Hi/Lo Speed)
- Alarm:
- 77dBA Low Voltage and 77dBA Low Flow
- 77dBA Low Voltage and 77dBA Low Flow
- 77dBA Low Voltage and 77dBA Low Flow
- 77dBA Low Voltage and 77dBA Low Flow
- Battery life:
- 4 - 10 h
- 4 - 10 h
- 4 - 10 h
- 4 - 10 h
- Battery type:
- Lithium Ion Polymer
- Lithium Ion Polymer
- Lithium Ion Polymer
- Lithium Ion Polymer
- Motor:
- Brushless
- Brushless
- Brushless
- Brushless