SOLA™ SPE Cartridges, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
SOLA products are available in a 10 mg/1 ml or 30 mg/3 ml cartridge format.
- Higher levels of reproducibility
- Higher levels of extract cleanliness
- Reduced solvent requirements
- Confidence in results
They provide highly reproducible, clean and consistent sample extracts for bioanalysis by utilizing the unique award winning fritless SPE technology. The enhanced performance of SOLA cartridges gives you greater confidence in your analytical results and lowers operating costs - without compromising ease of use or requiring complex method development.
Customers Who Bought This Also Bought