HyperSep™ Cyano SPE Cartridges
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Solid phase extraction cartridges with medium polarity sorbent, ideal for the isolation of polar compounds from non polar matrices. Optimized for the retention of polar compounds from non-polar matrices.
- Less retentive than silica or diol
- Used for retention of polar compounds from hexane and oils
- Suited for reversed phase extraction of moderately polar compounds
HyperSep-96 system incorporates traditional solid phase extraction sorbents into a 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customisable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing.
Packaging: HyperSep-96 are available either in packs of 100 individual wells or in pre-assembled plates.
Spécifications
- Catalog No:
- 10048-136
- 10048-134
- 10048-138
- 10048-130
- 10048-128
- 10048-126
- 10048-124
- Surface area:
- 525 m²/g
- 525 m²/g
- 525 m²/g
- 525 m²/g
- 525 m²/g
- 525 m²/g
- 525 m²/g