HyperSep™ Diol SPE Cartridges
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
A short alkyl chains with polar functional groups are ideal for the isolation of polar compounds.
- Normal phase extraction of polar compounds
- Purification of polar compounds
HyperSep-96 system incorporates traditional solid phase extraction sorbents into a 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customisable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing.
Packaging: HyperSep-96 are available either in packs of 100 individual wells or in pre-assembled plates.
Spécifications
- Catalog No:
- 10047-328
- 10047-326
- 10047-332
- 10047-330
- 10047-324
- 10047-322
- 10048-144
- 10048-142
- 10048-146
- Surface area:
- 470-530 m²/g
- 470-530 m²/g
- 470-530 m²/g
- 470-530 m²/g
- 470-530 m²/g
- 470-530 m²/g
- 470-530 m²/g
- 470-530 m²/g
- 470-530 m²/g