HyperSep™ SCX Strong Cation Exchanger SPE Cartridges
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
A strong cation sorbent for extraction of charged basic compounds.
Spécifications
- Catalog No:
- 10047-284
- 10047-372
- 10047-304
- 10047-370
- 10047-288
- 10047-286
- 10047-318
- 10048-078
- 10047-290
- 10048-076
- 10048-080
- Description:
- Surface area:
- 525 m²/g
- 525 m²/g
- 525 m²/g
- 525 m²/g
- 525 m²/g
- 525 m²/g
- 525 m²/g
- 525 m²/g
- 525 m²/g
- 525 m²/g
- 525 m²/g