HyperSep™ SAX Strong Anion Exchanger SPE Cartridges
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Strong anion exchange sorbent for extraction of weak acids.
Spécifications
- Catalog No:
- 10047-306
- 10048-070
- 10047-282
- 10047-234
- 10048-074
- 10048-072
- 10047-320
- 10047-280
- 10047-278
- Surface area:
- 481 m²/g
- 481 m²/g
- 481 m²/g
- 481 m²/g
- 481 m²/g
- 481 m²/g
- 481 m²/g
- 481 m²/g
- 481 m²/g