HyperSep™ Florisil SPE Cartridges
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Ideal for the isolation of polar compounds from non-polar matrices.
Customers Who Bought This Also Bought
Ideal for the isolation of polar compounds from non-polar matrices.