HyperSep™ Silica SPE Cartridges
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
A polar sorbent primarily used to retain analytes in non-polar matrices.
Spécifications
- Catalog No:
- 10048-068
- 10047-232
- 10047-276
- 10047-364
- 10047-230
- 10047-274
- 10047-296
- 10047-362
- 10047-368
- 10048-066
- 10047-366
- 10048-064
- 10047-272
- 10047-360
- Description:
- Surface area:
- 530 m²/g
- 530 m²/g
- 530 m²/g
- 530 m²/g
- 530 m²/g
- 530 m²/g
- 530 m²/g
- 530 m²/g
- 530 m²/g
- 530 m²/g
- 530 m²/g
- 530 m²/g
- 530 m²/g
- 530 m²/g