HyperSep™ C8 SPE Cartridges
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
HyperSep™ C8 cartridges, less retentive alternative to C18 for polar and nonpolar compounds.
- Well suited for method requiring less retention than C18
- For moderately polar analytes
- Potential for polar interactions greater than with C18
HyperSep-96 system incorporates traditional solid phase extraction sorbents into a 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing.
Packaging: HyperSep-96 are available either in packs of 100 individual wells or in pre-assembled plates.
Spécifications
- Catalog No:
- 10047-254
- 10047-298
- 10047-342
- 10047-252
- 10047-258
- 10047-346
- 10047-256
- 10047-228
- Surface area:
- 481 m²/g
- 481 m²/g
- 481 m²/g
- 481 m²/g
- 481 m²/g
- 481 m²/g
- 481 m²/g
- 481 m²/g