HyperSep™ C18 SPE Cartridges
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Feature a highly retentive alkyl-bonded phase for non-polar to moderately polar compounds.
Spécifications
- Catalog No:
- 10048-062
- 10047-340
- 10048-060
- 10048-152
- 10048-054
- 10047-354
- 10047-250
- 10047-338
- 10048-058
- 10047-336
- 10048-056
- 10047-238
- Surface area:
- 490 m²/g
- 490 m²/g
- 490 m²/g
- 490 m²/g
- 490 m²/g
- 490 m²/g
- 490 m²/g
- 490 m²/g
- 490 m²/g
- 490 m²/g
- 490 m²/g
- 490 m²/g