HyperSep™ Phenyl SPE Cartridges
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Solid phase extraction cartridges, HyperSep™ phenyl offer alternative selectivity for retention of basic compounds. They are used for extraction of aromatic compounds and extraction of basic analytes.
- Commonly employed for non-polar interactions
- The electron density of aromatic ring provides different selectivity than other non-polar sorbents
HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customisable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing.
Packaging: HyperSep-96 are available either in packs of 100 individual wells or in pre-assembled plates.
Spécifications
- Catalog No:
- 10047-350
- 10047-348
- 10047-310
- 10047-352
- 10047-246
- 10047-244
- 10047-248
- 10047-312
- 10047-242
- Surface area:
- 481 m²/g
- 481 m²/g
- 481 m²/g
- 481 m²/g
- 481 m²/g
- 481 m²/g
- 481 m²/g
- 481 m²/g
- 481 m²/g