HyperSep™ Retain PEP SPE Cartridges
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Versatile polymeric material for retention of polar and non-polar analytes.
- High and consistent recoveries
- Fast and easy sample preparation and method development
- pH stable zero to 14
HyperSep™ Retain™ PEP offers reproducible and reliable sample preparation through excellent batch-to-batch reproducibility. Many of the problems associated with traditional silica-based materials, such as inconsistent recoveries due to sorbent drying, are eliminated when using HyperSep™ Retain™ PEP. This can lead to fast sample preparation and method development.
Spécifications
- Catalog No:
- 10047-172
- 10047-164
- 10047-186
- 10047-170
- 10047-192
- 10047-168
- 10047-190
- 10047-180
- 10047-178
- 10047-182
- 10047-176
- 10047-174
- Surface area:
- 550-750 m²/g
- 550-750 m²/g
- 550-750 m²/g
- 550-750 m²/g
- 550-750 m²/g
- 550-750 m²/g
- 550-750 m²/g
- 550-750 m²/g
- 550-750 m²/g
- 550-750 m²/g
- 550-750 m²/g
- 550-750 m²/g