Omni Tissue Master 125 Handheld Homogenizer, Omni International, Inc.
Fournisseur: OMNI INTERNATIONAL
Certificats
A propos de cet article
The economically priced Tissue Master 125 is well suited for customrs who homogenize with little variation in sample size
The Tissue Master is lightweight enough to be hand-held, yet can also be mounted to a motor stand for lengthier processing.
Note: Omni Tip™ Plastic Probes are not compatible with the Tissue Master homogenizer.
The Omni Tissue Master includes one or more stainless steel generator probes that can handle a wide variety of sample types, making it a great choice for laboratories with many applications and a limited budget.
Certifications: CE compliant.
Ordering information: Includes 125 watt Tissue Master motor, stainless steel generator probes, 1-year warranty, and instruction manual.
Spécifications
- Catalog No:
- 10046-868
- 10046-866
- Sound level:
- \<72 db
- <72 db