VWR® Draw Stations
# de catalogue 10031-826
Fournisseur: VWR International
Spécifications
- Description:Phlebotomy Draw Station
- Cat. no.:10031-826
- L×W×H:15x13x3"
Spécifications
A propos de cet article
Keep all your supplies close at hand and organize small supplies with several compartments.
- Quick and convenient storage
- Clear plastic fabrication for easily identifying contents
This includes 2 tube racks for 13mm tubes and 17mm tubes.
Customers Who Bought This Also Bought