VWR® Acrylic Dispensing Bins
Fournisseur: VWR International
Certificats
A propos de cet article
The flip top lids on the top and on the front of these general use dispensing bins will keep your supplies clean and dust-free.
- Acrylic fabrication
- Can be easily opened with one hand
Made from premium acrylic, they’re available in three sizes. The lid is designed to have a slight overhang so you can lift it and pick up the items inside with one hand. Holds tube caps, cotton balls, pipette tips, analyzer cups—the possibilities are endless.
Ordering information: Accessories not included.
Customers Who Bought This Also Bought