Spécifications
- Description:BioCulture Calibration Head
- Cat. no.:102681-124
- Supplier No.:65224-50
Spécifications
A propos de cet article
The BioCulture Calibration Head is A NIST traceable venture calibrator with 1% accuracy that allow users to calibrate the flow of the BioCulture pump themselves without having to send the pump out for service of calibration.
- Allows flow calibration at flow rates between 30 – 120 LPM
- Designed for use with impeller fan type pumps like the BioCulture™, which require calibration devices with very little backpressure resistance