Spécifications
- Description:Dispensing Slide
- Cat. no.:102093-780
- Supplier No.:61727-08
Spécifications
A propos de cet article
These thin transparent polyvinyl slides serve as a surface to precisely orient the tissue into the desired position and as a means to accurately transfer tissue to the embedding well floor.
Tissue is applied face down to the end of the dispensing slide where it can be visualized from below and manipulated into position. The face that is visible will be laid onto the well floor and will ultimately be the embedded surface to be sectioned.