A propos de cet article
The DispoCut™ Disposable Dissecting Board is strong yet inexpensive enough to throw away. It is especially developed to provide a clean, safer and more efficient way of handling infectious tissue specimens for the pathologist.
- Can be used on both sides: great money saving feature
- Printed with helpful imperial and metric dimensional scales in inches, centimeters and millimeters
- Available in three sizes to accommodate small biopsies to large gross anatomy procedures
DispoCut™ is extrusion made using a copolymer resin in order to increase impact and high and low temperature performance. Copolymer resins are also used because they retain the ability to be flexed an unlimited number of times without breaking. Chemically, DispoCut is inert. At regular temperatures most oils, solvents and water have no effect, allowing it to perform under adverse weather conditions or as a product component exposed to harsh chemicals.
Each surface has many conveniently located rulers including seven dimensional circles from 4 to 16 mm in diameter and five larger ones from 2 to 6 cm in diameter. Surface will not dull knifes. Perfect for use with dissecting pins. All sizes of DispoCut™ Disposable Dissecting Boards are ideal for pathology labs, medical schools, classroom dissections, forensic labs, medical research labs, pharmaceutical labs, surgery and more. Temperature performance range: -17 °F to 160 °F, Melting point: 162 °C, 324 °F.
Sterilization: DispoCut™ may be wiped down with alcohol with no effect on the material. There are many types of autoclaves on the market. DispoCut™ is being autoclaved (in excess of 3 years) in many situations. There are some extreme settings which will be damaging and we suggest a trial to determine performance in your equipment. Ethylene oxide gas sterilization has no effect of an kind on DispoCut™ and may be repeated any number of times.
Each surface has many conveniently located rulers including seven dimensional circles from 4 to 16 mm in diameter and five larger ones from 2 to 6 cm in diameter. Surface will not dull knifes. Perfect for use with dissecting pins. All sizes of DispoCut™ Disposable Dissecting Boards are ideal for pathology labs, medical schools, classroom dissections, forensic labs, medical research labs, pharmaceutical labs, surgery and more. Temperature performance range: -17 °F to 160 °F, Melting point: 162 °C, 324 °F.
Sterilization: DispoCut™ may be wiped down with alcohol with no effect on the material. There are many types of autoclaves on the market. DispoCut™ is being autoclaved (in excess of 3 years) in many situations. There are some extreme settings which will be damaging and we suggest a trial to determine performance in your equipment. Ethylene oxide gas sterilization has no effect of an kind on DispoCut™ and may be repeated any number of times.
Copolymer resins are also used because they retain the ability to be flexed an unlimited number of times without breaking. Chemically, DispoCut is inert. At regular temperatures most oils, solvents and water have no effect, allowing it to perform under adverse weather conditions or as a product component exposed to harsh chemicals.