About this item
KODAK® Safelight filters are scientifically designed filters for safelight lamps.
OC/light amber - For contact and enlarging papers. Available in 51/2" Ø; 31/4×43/4"; 5×7"; 8×10" and 10×12".
1/red - For some blue-sensitive materials, most phototypesetting materials, KODAGRAPH Projection, and some KODAK LINAGRAPH papers. Available in 51/2" Ø; 5×7"; 8×10" and 10×12".
1A/light red - For slow orthochromatic materials. KODALITH and KODAGRAPH orthochromatic materials, and high resolution plates. Available in 51/2" Ø; 5×7"; 8×10" and 10×12".
2/dark red - For fast orthochromatic materials, some green-sensitive x-ray films, KODAK EKTALINE Papers, and KODAK orthochromatic LINAGRAPH papers. Available in 51/2" Ø; 8×10" and 10×12".
3/dark green - For some panchromatic materials. Available in 51/2" Ø; 8×10" and 10×12".
8/dark yellow - For some EASTMAN color Print and intermediate Films. Available in 10×12".
10/dark amber - For KODAK VERICOLOR slide film 5072, KODAK VERICOLOR print film 4111 (ESTAR Thick Base) KODAK PANALURE* Papers. Available only in 8×10".
13/amber - For all KODAK EKTACOLOR*, all KODAK PANALURE, and KODAK RESISTO rapid pan papers. Available in 51/2" Ø; 8×10" and 10×12".
GB X -2 - For all blue- and most green-sensitive medical and dental x-ray films. Available in 51/2" Ø; 31/4×43/4"; 5×7"; 8×10" and 10×12".
*Requires reduced bulb wattage.
It provides maximum safe illumination, when used as directed, plus offers protection from white light when using sensitized materials.