A propos de cet article
Used for working comfortably with solid state media.
- The Petri Dish Stand: Prevent Fatigues with the 30 angle creates a more comfortable work surface for streaking, transferring, and counting
- Has unique design, rear slot accepts all PetriOrienters to create a custom layout
The Petri Orienter: Quickly creates a consistent & area of reference on your petri dish. Sharp characters and lines allow for fast and easy position recognition. PetriOrienters are available in several layouts of grids or pie sectors to accommodate colonies of any size.