Spécifications
- Color:White
- Description:Bake Tough-Tags
- Supplier No.:77540-W
Spécifications
A propos de cet article
Ideal use with oven sterilization. Resists oven temperatures up to 180 °C, autoclavable, boilable and solvent resistant and heating to inactivate RNase, DNase and other enzymes.
- Adhere to any surface
- Bake Tags withstand cryogenic freezer storage conditions (liquid nitrogen)
- Withstand boiling water baths without experiencing any problems
Unlike conventional labels that become brittle, darken and fall off bottles, beakers, pipet canisters and other containers in the oven, bake tags resist most organic solvents and caustic agents, e.g. alcohols, most aromatic and some chlorinated solvents as well as acids and alkalis.