Spécifications
- Description:Mold Polyethylene
- Size:19x13 mm
- Supplier No.:70177-19
Spécifications
A propos de cet article
Molded from polyethylene, three popular sizes are available to meet your needs.
These Individual Molds can be used with EBH2 Block Holders, Aluminum Embedding Tray is available with 6-position to hold these Individual Mold during polymerization. Comes with anodized blue and natural color. Suitable for JB-4®, JB-4-Plus®, Immuno-Bed, LR White, LR Gold® or Paraffin embedding.