Cryogenic Grid Box, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
These specimen grid boxes are for storing or transferring cryogenic TEM specimen grids.
- Four diamond shaped slots
- Non-static clear cover shield held in place with stainless steel screw, which is tapped in the center of the box
- Box only fits 71166-20 (Grid box square with lid)
- Available with lid, pin-type lid, or without lid
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.