ContactUsLinkComponent
Cassette, PTFE Filters, 37 mm, Electron Microscopy Sciences
Cassette, PTFE Filters, 37 mm, Electron Microscopy Sciences
# de catalogue 102681-114
Air Sample Filter PTFE Zl 5 50/Pk
Cassette, PTFE Filters, 37 mm, Electron Microscopy Sciences
# de catalogue 102681-114
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Pore size:
    5.0 µm
  • Description:
    37 mm Cassette, 3 Piece, PTFE Filters Unsupport, (Zylon)
  • Cat. no.:
    102681-114
  • Supplier No.:
    65222-35
Spécifications
A propos de cet article
The 37 mm Air Sampling Cassette without 'PTFE' Filters are designed to meet all NIOSH, OSHA, and EPA requirement for air sampling standards.
All 37mm cassette housings are manufactured in house and are available in both two and three piece configurations. All 37mm cassettes are individually assembled and preloaded under stringent quality assurance standards with only the finest quality membrane and support pads.
Customers Who Bought This Also Bought