About this item
Reagent reservoir cooler are used to keep the reagent at ~5°C for approximately three hours
This cooler has frozen gel contained in a sealed molded cavity which extends safe working time for temperature sensitive reagents. Each 100 mL Reservoir fits perfectly and snuggly onto the Cooler ensuring uniform cooling. The 100 mL reservoirs are compatible with RESE-1000, -1050 and -1100 only.