Tough-Tag Color Coded Labels, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
Color coded tough tags will not come off in rotors, cyclers or racks.
- Ideal for use on 1.5 ml tubes
- Spot size is 0.44" (11 mm)
They are resistant to extreme temperatures and the matte surface is suitable for pencil, pen or marker.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.