Spécifications
- Description:Embedding Mold, Dykstra
- Cat. no.:102091-162
- Supplier No.:70907
Spécifications
A propos de cet article
This mold produces blocks of the best dimension to be held securely by Reichert/Leica Ultracut stepped microtome chucks.
EMS Embedding Mold is a new mold designed with Michael J. Dykstra Ph.D. from North Carolina State University Department of Veterinary Medicine. There are 24 cavities and the depth of each cavity is 4 mm which will prevent breakage of the blocks during razor blade trimming. Mold Measurements: 85×77 mm (3³/₈×3"). Cavity Measurements: 15×7×4 mm.