Richter Anti-Vibration Pad, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
Richter Anti-Vibration Pad inexpensively dampens vibrations from lab equipment and creates a stable surface that will keep equipment in place.
- Fits a variety of bench-top vortexes, shakers, rockers and microscopes
- Small, lightweight design makes transportation easy
- Pads can be stacked for heavier units or laid side-by-side for larger equipment
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.