96-Well and 384-Well Color Orienters, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
Fits most standard flat and round bottom 96-well and 384-well plates.
- Bright colors make it easy to distinguish between columns and rows
- Ideal for use with The Pipetter Guide for efficient and stress-free identification of rows and columns
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.