ContactUsLinkComponent
EMS Tissue Capture Pen, Electron Microscopy Sciences
EMS Tissue Capture Pen, Electron Microscopy Sciences
Catalog # 102097-464
CAS Number:  
Ems Tissue Capture Pen
EMS Tissue Capture Pen, Electron Microscopy Sciences
Catalog # 102097-464
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Tissue Capture Pen
  • Cat. no.:
    102097-464
  • Supplier No.:
    71314-10
Specifications
About this item
With one swift application of the EMS Tissue Capture Pen, you are able to change your plain glass slide into a coated one.
  • The EMS Tissue Capture Pen is a coating pen with a wide point (18 mm) for fixing various tissue sections (paraffin embedded, frozen and formalin sections, etc.) The unique pen is unaffected by heat up to 120°C so it offers secure and firm adhesion of the tissue section. It may be used in Microwave staining, immunostaining, in situ, and more. One application is all that is needed, and after it is applied, allow 1-2 minutes for drying. The EMS Tissue Capture may be used in conjunction with our Pap-Pen ( 71310) and will allow for easy placement of adherent as well as nonadherent cells on a glass slide. Immunostaining/ISH can be performed by applying a few drops of tissue suspension or liquid media onto the glass slide which is circled by the Pap-Pen, and then your section can be placed.
The EMS Tissue Capture Pen is a coating pen with a wide point (18 mm) for fixing various tissue sections (paraffin embedded, frozen and formalin sections, etc.) The unique pen is unaffected by heat up to 120°C so it offers secure and firm adhesion of the tissue section. It may be used in Microwave staining, immunostaining, in situ, and more. One application is all that is needed, and after it is applied, allow 1-2 minutes for drying. The EMS Tissue Capture may be used in conjunction with our Pap-Pen ( 71310) and will allow for easy placement of adherent as well as nonadherent cells on a glass slide. Immunostaining/ISH can be performed by applying a few drops of tissue suspension or liquid media onto the glass slide which is circled by the Pap-Pen, and then your section can be placed.
Customers Who Bought This Also Bought