Petri Dish Labels, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
PetriStickers™ are ideally sized stickers for labeling petri dishes.
- Label petri dishes quickly, neatly and reliably
- Labels remain firmly intact in temperatures from 4 to 42 °C
Instantly form an imprint petri dish for quick reference then can remove if need.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.