Breathable Microtube Membranes, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
Ideal for cell growth and freeze drying.
- Permits consistent gas exhange
- Easy microtube cap closure
- Will not bind in centrifuge rotors
- Easy-to-use finger tab for quick liner removal
- Pre-cut to fit a variety of popular tube sizes
- Sterile and free of cytotoxins
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.