CoolSafe Cool Bricks, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
Each Cool Brick allows up to 10 hours of benchtop temperature stability.
Two temperature zones are available: 1°C and -10°C.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.