Micro Spoon Heyman Type, Electron Microscopy Sciences
Fournisseur: Avantor
Certificats
A propos de cet article
A Heyman type micro spatula made from nickel stainless steel with a glazed finish.
- Wire Ø - 2.62 mm (0.103")
- Overall length: 159 mm (6¹/₄")
- Flat end: 31.35 mm (1¹/₄") long
The spoon end: 12.5 mm (½") long × 3 mm (¹/₈") wide. V-Shaped formed other end: ½" long.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.