Spécifications
- Description:Sample Storage Set, Sk-3
- Supplier No.:64269
Spécifications
A propos de cet article
Most economical, straight-sided, flat bottom, glass shell vial with high quality cork closures. Ideally suited for storing small parts, geological specimens, soil and many other types of samples.
Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card. Alpha-numerical indexing system allows quick identification and location of stored items. Vial size 3 dr (10.4 ml), 12 Ø×114 mm H, arrangement 5×20. Tray size 11½×3× 5".