Round Cover Slips – German Glass, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
These coverglass which are made from German glass and can be used for growing and culturing cells that normally have poor adhesion to plastic or regular glass surfaces.
- 5mm size will fit inside the 96 well plates
- Enough room to pick up 5 mm from the bottom of the well with forceps
- 8 mm size fits inside the 24 well plates
- Available with #1.5 (0.16 – 0.19 mm) thickness
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.