EZ Crimper™, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
Adjustable stoppers on these crimpers allow the user to determine the tightness of the seal while ensuring consistent crimping. Ergonomic, PVC grip and spring assembly reduces hand fatigue.
- Tapered jaws allow for crimping of the vials in the racks
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.