A propos de cet article
Tough-Tags® labels are mainly used for microcentrifuge tubes and other containers. These labels are made of chemically inert polyester sheets which resist most organic solvents and caustic agents.
- Adhere to most plastics, glass and metals without cracking, peeling or degrading
- Temperature resistance: −40 to 121 °C
Label sheets are specially top-coated to readily accept laser printing. Stay-flat sheets will not jam in laser printers. Tubes labeled with Tough-Tags® are easily slide in and out of centrifuge rotors without binding.