Aluminum Oxide Abrasive Film Discs, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
Alumina is widely used in sample grinding and polishing. Our Aluminum Oxide Abrasive film with a PSA back or a plain back are ready to use in all sample polishing applications.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.