ContactUsLinkComponent
UltraBed Kit, Electron Microscopy Sciences
UltraBed Kit, Electron Microscopy Sciences
Catalog # 101410-644
Minority or Woman-Owned Business Enterprise
CAS Number:  
ULTRABED KIT
UltraBed Kit, Electron Microscopy Sciences
Catalog # 101410-644
Minority or Woman-Owned Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    UltraBed Low Viscosity Kit
  • Size:
    2 x 100 ml
  • Cat. no.:
    101410-644
  • Supplier No.:
    14310
Specifications
About this item
The kit consists of two components, equal amount, very low viscosity, (~65 cps), in convenient dispenser top bottles. To prepare the embedding resin, just mix equal amounts of the two components. The resin readily infiltrates into specimen and polymerizes to a clear, hard block overnight at 60°C.
  • A Newly Introduced Low Viscosity Epoxy Embedding Kit
  • Mixed resin can be used for both infiltration and embedding
  • All components are shipped together as Non- Hazardous substances
Ordering information: Kit comes with 100 mL each of component A and component B.
Customers Who Bought This Also Bought