Manomark Pen All Surface Marker, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
Writes on all plastic, glass, metal, wool, paper and cloth, including "PTFE", fluoropolymer resins
- With its precision seal valve the pens life is preserved and as well it allows for a smooth flow of ink even if it’s left uncapped
- Once dry, the solvent base ink is temperature resistant from 0 to 155 °C (32 to 310 °F)
Markings are insoluble in water or hydrocarbons, resist many solvents, and are easily removed with alcohol.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.